Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ropivacaine hydrochloride monohydrate | 132112-35-7 | 99.9% | 1 ML
SDP

Catalog No. 5000408620
Encompass_Preferred

Ropivacaine hydrochloride monohydrate is a potent sodium channel blocker that inhibits impulse conduction by reversibly blocking sodium ion influx in nerve fibers. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1. This product is extensively used for regional anesthesia and the management of neuropathic pain in vivo.

  • Potent sodium channel blocker
  • Inhibits impulse conduction by blocking sodium ion influx
  • Inhibitor of K2P (two-pore domain potassium channel) TREK-1
  • Purity of 99.93%
  • Appearance: white to off-white solid
  • Recommended storage for solid: 4°C, sealed, away from moisture
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-08620 Supplier Medchemexpress LLC Supplier No. HYB0563A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ropivacaine hydrochloride monohydrate is a potent sodium channel blocker that inhibits impulse conduction by reversibly blocking sodium ion influx in nerve fibers. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1. This product is extensively used for regional anesthesia and the management of neuropathic pain in vivo.

  • Potent sodium channel blocker
  • Inhibits impulse conduction by blocking sodium ion influx
  • Inhibitor of K2P (two-pore domain potassium channel) TREK-1
  • Purity of 99.93%
  • Appearance: white to off-white solid
  • Recommended storage for solid: 4°C, sealed, away from moisture
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.