Learn More
Apexbio Technology LLC ROPIVACAINE-MESILATE-50MG
SDP
Catalog No. 5000568834
Encompass_Preferred
Ropivacaine Mesilate (CAS No 854056-07-8) is a local anesthetic of the amino amide type commonly studied in biomedical research for its nerve-blocking properties Originating from chemical modifications of pipecoloxylidide it targets sodium ion channels inhibiting nerve signal transmission which is central to its mechanism of action In vitro it exhibits activity in the nanomolar to low micromolar range Research applications include nerve cell models and various animal studies to explore its efficacy and safety profile particularly in drug screening and pain management Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68834 Supplier Apexbio Technology LLC Supplier No. C650750
Ropivacaine Mesilate (CAS No 854056-07-8) is a local anesthetic of the amino amide type commonly studied in biomedical research for its nerve-blocking properties Originating from chemical modifications of pipecoloxylidide it targets sodium ion channels inhibiting nerve signal transmission which is central to its mechanism of action In vitro it exhibits activity in the nanomolar to low micromolar range Research applications include nerve cell models and various animal studies to explore its efficacy and safety profile particularly in drug screening and pain management Experimental concentrations typically depend on the specific research design