Learn More
Cayman Chemical RopIvacaInhydrochlorId 50mg
SDP
Catalog No. 502995399
Encompass_Preferred
A potent and reversible sodium channel blocker; induces complete impairment of proprioception, motor function, and nociception in the hindleg of rats when 100 μL of an 8 mM solution is injected percutaneously into the sciatic nerve; depresses myocardial contractile force in isolated rat hearts less potently than (±)-ropivacaine, as well as (–)- and (±)-bupivacaine
Catalog No. 50-299-5399 Supplier Cayman Chemical Supplier No. 2142250 MG
A potent and reversible sodium channel blocker; induces complete impairment of proprioception, motor function, and nociception in the hindleg of rats when 100 μL of an 8 mM solution is injected percutaneously into the sciatic nerve; depresses myocardial contractile force in isolated rat hearts less potently than (±)-ropivacaine, as well as (–)- and (±)-bupivacaine