Learn More
Cayman Chemical (–)-Ropivacaine (hydrochloride hydrate), Purity: ≥98%, CAS: 132112-35-7, 100 mg
SDP
Catalog No. 502994030
Encompass_Preferred
A potent and reversible sodium channel blocker; induces complete impairment of proprioception, motor function, and nociception in the hindleg of rats when 100 μL of an 8 mM solution is injected percutaneously into the sciatic nerve; depresses myocardial contractile force in isolated rat hearts less potently than (±)-ropivacaine, as well as (–)- and (±)-bupivacaine
Catalog No. 50-299-4030 Supplier Cayman Chemical Supplier No. 21422100 MG
A potent and reversible sodium channel blocker; induces complete impairment of proprioception, motor function, and nociception in the hindleg of rats when 100 μL of an 8 mM solution is injected percutaneously into the sciatic nerve; depresses myocardial contractile force in isolated rat hearts less potently than (±)-ropivacaine, as well as (–)- and (±)-bupivacaine