Learn More
Aviva Systems Biology Corp ROPN1L Antibody - N-terminal region (ARP60573_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60573_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ROPN1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ROPN1L. Alternate names: ASP, FLJ23003, FLJ25776, RSPH11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-088