Learn More
Phenomenex Inc roQ QuEChERS dSPE Kit - 15mL CT
Catalog No. 502398007 Shop All Phenomenex Products
Encompass_Preferred
roQ QuEChERS dSPE Kit - 15mL CT 1200mg MgSO4,400mg PSA,400mg C18E,400mg GCB
Catalog No. 50-239-8007 Supplier Phenomenex Inc Supplier No. KS09518
roQ QuEChERS dSPE Kit - 15mL CT 1200mg MgSO4,400mg PSA,400mg C18E,400mg GCB