Learn More
Phenomenex Inc roQ QuEChERS dSPE Kit - 15mL CT
Catalog No. 502399504 Shop All Phenomenex Products
Encompass_Preferred
roQ QuEChERS dSPE Kit - 15mL CT 750mg MgSO4, 125mg PSA, 37.5mg GCB
Catalog No. 50-239-9504 Supplier Phenomenex Inc Supplier No. KS08930
roQ QuEChERS dSPE Kit - 15mL CT 750mg MgSO4, 125mg PSA, 37.5mg GCB