Learn More
Phenomenex Inc 1200mg MgSO4, 400mg PSA, 400mg C18E
Catalog No. 502401022 Shop All Phenomenex Products
Encompass_Preferred
roQ™ QuEChERS dSPE Kit - 15mL CT KS0-9516
Catalog No. 50-240-1022 Supplier Phenomenex Inc Supplier No. KS09516
roQ™ QuEChERS dSPE Kit - 15mL CT KS0-9516