Learn More
Phenomenex Inc 900mg MgSO4, 150mg PSA, 45mg GCB
Catalog No. 502406747 Shop All Phenomenex Products
Encompass_Preferred
roQ™ QuEChERS dSPE Kit - 15mL CT KS0-9510
Catalog No. 50-240-6747 Supplier Phenomenex Inc Supplier No. KS09510
roQ™ QuEChERS dSPE Kit - 15mL CT KS0-9510