Learn More
Phenomenex Inc roQ QuEChERS dspe Kit - 15mL CT
Catalog No. 502397880 Shop All Phenomenex Products
Encompass_Preferred
roQ QuEChERS dspe Kit - 15mL CT 900mg MgSO4, 150mg PSA, 150mg C18E
Catalog No. 50-239-7880 Supplier Phenomenex Inc Supplier No. KS09508
roQ QuEChERS dspe Kit - 15mL CT 900mg MgSO4, 150mg PSA, 150mg C18E