Learn More
Phenomenex Inc 150mg MgSO4, 50mg PSA, 50mg GCB
Catalog No. 502406755 Shop All Phenomenex Products
Encompass_Preferred
roQ™ QuEChERS dSPE Kit - 2mL CT KS0-9513
Catalog No. 50-240-6755 Supplier Phenomenex Inc Supplier No. KS09513
roQ™ QuEChERS dSPE Kit - 2mL CT KS0-9513