Learn More
Phenomenex Inc 150mg MgSO4, 25mg PSA, 25mg C18E
Catalog No. 502396837 Shop All Phenomenex Products
Encompass_Preferred
roQ™ QuEChERS dSPE Kit - 2mL CT KS0-9504
Catalog No. 50-239-6837 Supplier Phenomenex Inc Supplier No. KS09504
roQ™ QuEChERS dSPE Kit - 2mL CT KS0-9504