Learn More
Phenomenex Inc roQ QuEChERS dSPE Kit - 2mL CT
Catalog No. 502399482 Shop All Phenomenex Products
Encompass_Preferred
roQ QuEChERS dSPE Kit - 2mL CT 150mg MgSO4, 25mg PSA, 2.5mg GCB
Catalog No. 50-239-9482 Supplier Phenomenex Inc Supplier No. KS09505
roQ QuEChERS dSPE Kit - 2mL CT 150mg MgSO4, 25mg PSA, 2.5mg GCB