Learn More
Phenomenex Inc 4.0g MgSO4, 1.0g NaCl, 1.0g SCTD, 0.5g SCDS
Supplier:  Phenomenex Inc KS08909NC
Encompass_Preferred
roQ™ QuEChERS Extraction Kit,EN Method-No Collar KS0-8909-NC
Catalog No. 50-239-7350