Learn More
Enzo Life Sciences Roquefortine C (0.5mg). CAS: 58735-64-1
SDP
Supplier:  Enzo Life Sciences ALX350342MC05
Encompass_Preferred
Potent neurotoxin produced by a diverse range of fungi, most notably Penicillium species. Inhibits growth of Gram-positive bacteria. Inhibits cytochrome p450. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8852