Learn More
AdipoGen Roquefortine C
SDP
Supplier:  AdipoGen BVT0425C500
Encompass_Preferred
Chemical. CAS 58735-64-1. Formula C22H23N5O2. MW 389.5. Isolated from Penicillium sp. Potent neurotoxin. Mycotoxin. Tremorgenic. Gram-positive bacteria growth inhibitor. Cytochrome p450 inhibitor. Lymphocyte proliferation inhibitor. Cytotoxic.
Catalog No. 50-256-1935