Learn More
Abcam Roquefortine C, Neuroto x in, 500UG
Catalog No. 502705859
Encompass_Preferred
MW 389.4 Da, Purity >98%. Mycotoxin. Potent neurotoxin. Inhibits Cytochrome p450. Inhibits lymphocyte proliferation. Penicillin (ab145634, ab143435) metabolite. Shows tremorgenic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5859 Supplier Abcam Supplier No. AB144345500UG
MW 389.4 Da, Purity >98%. Mycotoxin. Potent neurotoxin. Inhibits Cytochrome p450. Inhibits lymphocyte proliferation. Penicillin (ab145634, ab143435) metabolite. Shows tremorgenic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement