Learn More
Medchemexpress LLC Roquinimex | 84088-42-6 | MFCD00866331 | 99.7% | 308.3 g/mol | C18H16N2O3 | 5 MG
SDP
Catalog No. 5000222187
Encompass_Preferred
Roquinimex (linomide; CAS 84088-42-6) is a quinoline-3-carboxamide research chemical described as an immunostimulant and TNF receptor inhibitor. It is supplied as a high-purity analytical standard intended for research and analytical applications.
  • High purity (99.7%).
  • Suitable as an analytical standard for research applications.
  • Quinoline-3-carboxamide class immunostimulant and TNF receptor inhibitor.
  • Supplied in small, research-scale solid vials for easy handling.
  • Molecular formula C18H16N2O3; molecular weight 308.3 g/mol.
Catalog No. 50-002-22187 Supplier Medchemexpress LLC Supplier No. HY137435MG
Roquinimex (linomide; CAS 84088-42-6) is a quinoline-3-carboxamide research chemical described as an immunostimulant and TNF receptor inhibitor. It is supplied as a high-purity analytical standard intended for research and analytical applications.
  • High purity (99.7%).
  • Suitable as an analytical standard for research applications.
  • Quinoline-3-carboxamide class immunostimulant and TNF receptor inhibitor.
  • Supplied in small, research-scale solid vials for easy handling.
  • Molecular formula C18H16N2O3; molecular weight 308.3 g/mol.