Learn More
Medchemexpress LLC RORγt inverse agonist 13 | 2170477-75-3 | 98.1% | 513.29 | C23H17Cl2F3N2O4 | 5 MG
SDP
Catalog No. 5000262117
Encompass_Preferred
RORγt inverse agonist 13 (Compound 3i) is a potent, orally active, and selective inverse agonist of the RORγt nuclear receptor with a reported IC50 of 63.8 nM. The compound is supplied for research use and is available as solid and solution formulations with high analytical purity.
  • Potent inverse agonist of RORγt with IC50 of 63.8 nM.
  • Orally active and selective for RORγt.
  • High purity (98.1%) suitable for research applications.
  • Molecular weight 513.29; formula C23H17Cl2F3N2O4.
  • Available as solid (5-500 mg) and as DMSO solutions for convenient dosing.
Catalog No. 50-002-62117 Supplier Medchemexpress LLC Supplier No. HY1313385MG
RORγt inverse agonist 13 (Compound 3i) is a potent, orally active, and selective inverse agonist of the RORγt nuclear receptor with a reported IC50 of 63.8 nM. The compound is supplied for research use and is available as solid and solution formulations with high analytical purity.
  • Potent inverse agonist of RORγt with IC50 of 63.8 nM.
  • Orally active and selective for RORγt.
  • High purity (98.1%) suitable for research applications.
  • Molecular weight 513.29; formula C23H17Cl2F3N2O4.
  • Available as solid (5-500 mg) and as DMSO solutions for convenient dosing.