Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RORγt inverse agonist 23 | 2230779-18-5 | 99.3% | 471.61 | 50 MG
SDP

Catalog No. 5000351962
Encompass_Preferred

RORγt inverse agonist 23 is a potent, selective, and orally available novel retinoic acid receptor-related orphan receptor γt inverse agonist.

  • Potent and selective RORγt inverse agonist
  • Orally available
  • Appearance: white to off-white solid
  • Molecular Formula: C26H33NO5S

Catalog No. 50-003-51962 Supplier Medchemexpress LLC Supplier No. HY13984750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RORγt inverse agonist 23 is a potent, selective, and orally available novel retinoic acid receptor-related orphan receptor γt inverse agonist.

  • Potent and selective RORγt inverse agonist
  • Orally available
  • Appearance: white to off-white solid
  • Molecular Formula: C26H33NO5S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.