Learn More
Medchemexpress LLC RORγt inverse agonist 23 | 2230779-18-5 | 99.3% | 471.61 g·mol⁻¹ | C26H33NO5S | 5 MG
SDP
Catalog No. 5000221162
Encompass_Preferred
RORγt inverse agonist 23 is a potent, selective, and orally available retinoic acid receptor-related orphan receptor γt (RORγt) inverse agonist for research use, used to probe RORγt-mediated signaling and evaluate pharmacological effects in preclinical models.
  • Potent and selective inverse agonist of RORγt.
  • Orally available in pharmacological models.
  • High purity (99.3%) suitable for biochemical and cellular assays.
  • Supplied as a white to off-white solid; soluble in DMSO.
  • Stable when stored sealed at recommended temperatures.
  • Offered in multiple mass formats and pre-made solution aliquots for convenience.
Catalog No. 50-002-21162 Supplier Medchemexpress LLC Supplier No. HY1398475MG
RORγt inverse agonist 23 is a potent, selective, and orally available retinoic acid receptor-related orphan receptor γt (RORγt) inverse agonist for research use, used to probe RORγt-mediated signaling and evaluate pharmacological effects in preclinical models.
  • Potent and selective inverse agonist of RORγt.
  • Orally available in pharmacological models.
  • High purity (99.3%) suitable for biochemical and cellular assays.
  • Supplied as a white to off-white solid; soluble in DMSO.
  • Stable when stored sealed at recommended temperatures.
  • Offered in multiple mass formats and pre-made solution aliquots for convenience.