Learn More
Medchemexpress LLC RORγt Inverse agonist 6 | 1887161-80-9 | 99.0% | 100 MG
SDP
Catalog No. 5000340850
Encompass_Preferred
RORγt Inverse agonist 6 (compound 43) is a RORγt inverse agonist for the study of Th17-driven autoimmune diseases. It suppresses IL-17A gene expression by IL-23 stimulation.
  • RORγt inverse agonist.
  • Used for studying Th17-driven autoimmune diseases.
  • Suppresses IL-17A gene expression.
Catalog No. 50-003-40850 Supplier Medchemexpress LLC Supplier No. HY130243100MG
RORγt Inverse agonist 6 (compound 43) is a RORγt inverse agonist for the study of Th17-driven autoimmune diseases. It suppresses IL-17A gene expression by IL-23 stimulation.
  • RORγt inverse agonist.
  • Used for studying Th17-driven autoimmune diseases.
  • Suppresses IL-17A gene expression.