Learn More
Medchemexpress LLC Rorγt inverse agonist 6 | 1887161-80-9 | MFCD32263437 | 98.97% | C28H29ClN6O5 | 10MG
SDP
Catalog No. 5000259729
Encompass_Preferred
RORγt Inverse agonist 6 is a research compound acting as an inverse agonist of the transcription factor RORγt. It is used in preclinical studies to investigate Th17-mediated inflammation and has been reported to suppress IL-17A expression following IL-23 stimulation in vivo.
  • Inverse agonist activity against RORγt suitable for mechanistic studies.
  • Demonstrated suppression of IL-17A expression in vivo.
  • Available as solid powder and as a ready-to-use DMSO solution for assay flexibility.
  • High purity (~98.97%) appropriate for biochemical and cellular assays.
  • Characterized by molecular formula C28H29ClN6O5 and molecular weight 565.02.
Catalog No. 50-002-59729 Supplier Medchemexpress LLC Supplier No. HY13024310MG
RORγt Inverse agonist 6 is a research compound acting as an inverse agonist of the transcription factor RORγt. It is used in preclinical studies to investigate Th17-mediated inflammation and has been reported to suppress IL-17A expression following IL-23 stimulation in vivo.
  • Inverse agonist activity against RORγt suitable for mechanistic studies.
  • Demonstrated suppression of IL-17A expression in vivo.
  • Available as solid powder and as a ready-to-use DMSO solution for assay flexibility.
  • High purity (~98.97%) appropriate for biochemical and cellular assays.
  • Characterized by molecular formula C28H29ClN6O5 and molecular weight 565.02.