Learn More
Aviva Systems Biology Corp ROR1 Antibody - N-terminal region (ARP63926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ROR1. It was validated on Western Blot. . Alternate names: MGC99659, NTRKR1, dJ537F10.1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-119