Learn More
ACROBiosystems ACROBiosystems Human ROR1 Protein, Fc Tag
Supplier:  ACROBiosystems RO1H5250200UG
Encompass_Preferred
Recombinant Protein;200UG;Human ROR1, Fc Tag (RO1-H5250) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.6 kDa. The protein migrates as 79-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ROR1, Fc Tag (Cat. No. RO1-H5250) at 2 μg/mL (100 μL/well) can bind Anti-Human ROR1 MAb with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-195-7129