Learn More
ACROBiosystems ACROBiosystems Biotinylated Human ROR1 Protein, Fc,Avitag™
Catalog No. 501957439
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human ROR1, Fc,Avitag (RO1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 70.4 kDa. As a result of Glycosylation, the protein migrates as 79-104 kDa under reducing (R) condition, and 158-208 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Anti-Human ROR1 MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human ROR1, Fc,Avitag (Cat. No. RO1-H82F4) with a linear range of 0.5-8 ng/mL (QC tested).
Catalog No. 50-195-7439 Supplier ACROBiosystems Supplier No. RO1H82F4200UG
Recombinant Protein;200UG;Biotinylated Human ROR1, Fc,Avitag (RO1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 70.4 kDa. As a result of Glycosylation, the protein migrates as 79-104 kDa under reducing (R) condition, and 158-208 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Anti-Human ROR1 MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human ROR1, Fc,Avitag (Cat. No. RO1-H82F4) with a linear range of 0.5-8 ng/mL (QC tested).