Learn More
Medchemexpress LLC ROR1 Human sf9 Hi 10ug
SDP
Catalog No. 5000180590
Encompass_Preferred
ROR1 protein s minimal kinase activity in vitro suggests an improbable role as a tyrosine kinase in vivo It serves as a receptor for WNT5A activating NFkB signaling and potentially inhibiting WNT3A signaling Notably ROR1 plays a vital role in the inner ear facilitating innervation of auditory hair cells by spiral ganglion neurons ROR1 Protein Human (sf9 His) is the recombinant human-derived ROR1 protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-80590 Supplier Medchemexpress LLC Supplier No. HYP7339010UG
ROR1 protein s minimal kinase activity in vitro suggests an improbable role as a tyrosine kinase in vivo It serves as a receptor for WNT5A activating NFkB signaling and potentially inhibiting WNT3A signaling Notably ROR1 plays a vital role in the inner ear facilitating innervation of auditory hair cells by spiral ganglion neurons ROR1 Protein Human (sf9 His) is the recombinant human-derived ROR1 protein expressed by Sf9 insect cells with C-His labeled tag