Learn More
Sino Biological Recombinant Human ROR1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 13968H08H1
Encompass_Preferred
A DNA sequence encoding the human ROR1 (NP_005003.2) (Met1-Glu403) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6350