Learn More
Sino Biological Recombinant Mouse ROR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 5A0666M08H100
Encompass_Preferred
A DNA sequence encoding the mouse ROR1 (NP_038873.2) (Met1-Glu403) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6487