Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human ROR1 Protein, Avi Tag (Avitag™);HEK293;25UG;RO1,ROR1,NTRKR1,ROR-1,NTRKR-1
Supplier: ACROBiosystems RO1H821Y25UG
Recombinant Protein;Biotinylated Human ROR1 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human ROR1, Avi Tag (RO1-H821y) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries an Avi tag at the C-terminus. The protein has a calculated MW of 45.2 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.