Learn More
ACROBiosystems Recombinant Protein;Mouse ROR1 Protein, Fc Tag;HEK293;1MG;RO1,ROR1,NTRKR1,ROR-1,NTRKR-1
Catalog No. 501658682
Encompass_Preferred
Recombinant Protein;Mouse ROR1 Protein, Fc Tag;1MG;Mouse ROR1, Fc Tag (RO1-M5250) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q9Z139).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.4 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8682 Supplier ACROBiosystems Supplier No. RO1M52501MG
Recombinant Protein;Mouse ROR1 Protein, Fc Tag;1MG;Mouse ROR1, Fc Tag (RO1-M5250) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q9Z139).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.4 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.