Learn More
ACROBiosystems Recombinant Protein;Mouse ROR1 Protein;HEK293;1MG;RO1,ROR1,NTRKR1,ROR-1,NTRKR-1
Supplier:  ACROBiosystems RO1M52211MG
Encompass_Preferred
Recombinant Protein;Mouse ROR1 Protein;1MG;Mouse ROR1, His Tag (RO1-M5221) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q9Z139).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8686