Learn More
ACROBiosystems Recombinant Protein;Rat ROR1 Protein;HEK293;1MG;RO1,ROR1,NTRKR1,ROR-1,NTRKR-1
Catalog No. 501658464
Encompass_Preferred
Recombinant Protein;Rat ROR1 Protein;1MG;Rat ROR1, His Tag (RO1-R5221) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # EDL97819).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 60-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8464 Supplier ACROBiosystems Supplier No. RO1R52211MG
Recombinant Protein;Rat ROR1 Protein;1MG;Rat ROR1, His Tag (RO1-R5221) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # EDL97819).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 60-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.