Learn More
ACROBiosystems Recombinant Protein;Human ROR1 (39-151, Ig-like domain) Protein;HEK293;50UG;RO1,ROR1,NTRKR1,ROR-1,NTRKR-1
Catalog No. 501658917
Encompass_Preferred
Recombinant Protein;Human ROR1 (39-151, Ig-like domain) Protein;50UG;Human ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession # Q01973).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.4 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8917 Supplier ACROBiosystems Supplier No. RO1H522150UG
Recombinant Protein;Human ROR1 (39-151, Ig-like domain) Protein;50UG;Human ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession # Q01973).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.4 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.