Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human ROR1 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;RO1,ROR1,NTRKR1,ROR-1,NTRKR-1
Supplier:  ACROBiosystems RO1H82E625UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human ROR1 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human ROR1, His Tag, Avi Tag (RO1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.7 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7716