Learn More
ACROBiosystems ACROBiosystems Mouse ROR2 / NTRKR2 Protein, Fc Tag
Catalog No. 501957282
Encompass_Preferred
Recombinant Protein;1MG;Mouse ROR2, Fc Tag (RO2-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # Q9Z138-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.6 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7282 Supplier ACROBiosystems Supplier No. RO2M52531MG
Recombinant Protein;1MG;Mouse ROR2, Fc Tag (RO2-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # Q9Z138-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.6 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.