Learn More
ACROBiosystems ACROBiosystems Human ROR2 / NTRKR2 Protein, His Tag
Supplier:  ACROBiosystems RO2H52E51MG
Encompass_Preferred
Recombinant Protein;1MG;Human ROR2, His Tag (RO2-H52E5) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # NP_004551.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7291