Learn More
ACROBiosystems ACROBiosystems Biotinylated Human ROR2 / NTRKR2 Protein, His,Avitag™
Catalog No. 501957203
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # A1L4F5-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7203 Supplier ACROBiosystems Supplier No. RO2H82E325UG
Recombinant Protein;25UG;Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # A1L4F5-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.