Learn More
Sino Biological Human ROR2 Protein (His & AVI Tag), Biotinylated 100ug
Supplier:  Sino Biological 16133H27HB100
Encompass_Preferred
A DNA sequence encoding the human ROR2 (NP_004551.2) (Met1-Gly403) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5370