Learn More
ACROBiosystems Recombinant Protein;Mouse ROR2 / NTRKR2 Protein, Fc Tag;HEK293;100UG;RO2,ROR2,NTRKR2,ROR-2,NTRKR-2
Supplier:  ACROBiosystems RO2M5253100UG
Encompass_Preferred
Recombinant Protein;Mouse ROR2 / NTRKR2 Protein, Fc Tag;100UG;Mouse ROR2, Fc Tag (RO2-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # Q9Z138-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.8 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8414