Learn More
ACROBiosystems Recombinant Protein;Human ROR2 / NTRKR2 Protein, Fc Tag;HEK293;1MG;RO2,ROR2,NTRKR2,ROR-2,NTRKR-2
Catalog No. 501658629
Encompass_Preferred
Recombinant Protein;Human ROR2 / NTRKR2 Protein, Fc Tag;1MG;Human ROR2, Fc Tag (RO2-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # AAI30523.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8629 Supplier ACROBiosystems Supplier No. RO2H52511MG
Recombinant Protein;Human ROR2 / NTRKR2 Protein, Fc Tag;1MG;Human ROR2, Fc Tag (RO2-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # AAI30523.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.