Learn More
Aviva Systems Biology Corp RORA Antibody - N-terminal region (P100794_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100794_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RORA. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RORA. Alternate names: ROR1, ROR2, ROR3, RZRA, NR1F1, RZR-ALPHAApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-62714