Learn More
Aviva Systems Biology Corp RORA Antibody - middle region (ARP45608_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45608_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RORA. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RORA. Alternate names: MGC119326, MGC119329, NR1F1, ROR1, ROR2, ROR3, RZRA, RZR-ALPHAApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59066