Learn More
Aviva Systems Biology Corp RORA Antibody - N-terminal region (ARP45642_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45642_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RORA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RORA. Alternate names: MGC119326, MGC119329, NR1F1, ROR1, ROR2, ROR3, RZRA, RZR-ALPHAApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59090