Learn More
Aviva Systems Biology Corp Rorb Antibody - N-terminal region (ARP38878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rorb. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-763