Learn More
Aviva Systems Biology Corp Rorb Antibody - N-terminal region (ARP45630_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rorb. It was validated on Western Blot. . Alternate names: MGC38728, Nr1f2, RZR-beta, RZRB, RorbetaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-285-210