Learn More
Aviva Systems Biology Corp RORC Antibody - N-terminal region (ARP59155_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59155_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RORC. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RORC. Alternate names: MGC129539, NR1F3, RORG, RZR-GAMMA, RZRG, TORApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-597