Learn More
Aviva Systems Biology Corp RORC Antibody - N-terminal region (ARP45622_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45622_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RORC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RORC. Alternate names: MGC129539, NR1F3, RORG, RZRG, TOR, RZR-GAMMAApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59075