Learn More
Aviva Systems Biology Corp RORC Antibody - middle region (ARP33243_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33243_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RORC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RORC. Alternate names: MGC129539, NR1F3, RORG, RZRG, TOR, RZR-GAMMAApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52897