Learn More
AdipoGen Roridin E
SDP
Supplier:  AdipoGen AGCN20176C250
Encompass_Preferred
Chemical. CAS 16891-85-3. Formula C29H38O5. MW 514.6. Isolated from fungus Trichoderma sp. Mycotoxin. Implicated in human and animal toxicosis. Potent cytotoxic and antiproliferative agent against cancer cell lines. Potent antimalarial agent. Antifungal, antibiotic, phototoxic and cytostatic agent. Antiviral against arenavirus Junin JUNV.
Catalog No. 50-256-2639